Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567631 Supplier Invitrogen™ Supplier No. PA567631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Receptor for netrin required for axon guidance. Mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Mediates apoptosis by activating DAPK1. In the absence of NTN1, activates DAPK1 by reducing its autoinhibitory phosphorylation at Ser-308 thereby increasing its catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UNC5B
Gene Accession No. Q8IZJ1, Q8K1S3
Gene Alias 6330415E02Rik; A630020F16; C. elegans; D10Bwg0792e; Netrin receptor UNC5B; p53RDL1; p53-regulated receptor for death and life protein 1; Protein unc-5 homolog 2; protein unc-5 homolog B; transmembrane receptor Unc5H2; unc5 homolog 2; unc-5 homolog 2; unc-5 homolog B; unc-5 homolog B (C. elegans); unc-5 netrin receptor B; Unc5b; UNC5H2; UNQ1883/PRO4326
Gene Symbols UNC5B
Host Species Rabbit
Immunogen Synthesized peptide derived from human UNC5B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107449, 219699
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.