Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114231 Supplier Invitrogen™ Supplier No. PA5114231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVSIEISRQQ VEELFGPEDY WCQCVAWSSA GTTKSRKAYV RIAYLRKTFE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin -like domains and 2 type I thrombospondin motifs in the extracellular region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Unc5c
Gene Accession No. O95185
Gene Alias B130051O18Rik; Netrin receptor UNC5C; protein unc-5 homolog 3; Protein unc-5 homolog C; Rcm; Rostral cerebellar malformation protein; unc5 (C.elegans homolog) c; unc5 homolog 3; UNC-5 homolog 3; unc-5 homolog C; unc-5 homolog C (C. elegans); unc-5 netrin receptor C; UNC5C; UNC5H3
Gene Symbols Unc5c
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Human UNC5C.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8633
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.