Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC5CL Antibody, Novus Biologicals™
SDP

Catalog No. p-5093692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224576SS 0.025 mg
NBP224576 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224576SS Supplier Novus Biologicals Supplier No. NBP224576SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC5CL Polyclonal specifically detects UNC5CL in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UNC5CL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias MGC34763, Protein unc-5 homolog C-like, unc-5 homolog C (C. elegans)-like, ZU5 and death domain containing, ZU5 and death domain-containing protein, ZUDUNC5C-like protein
Gene Symbols UNC5CL
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 49-100 and 500-561 of human UNC5CL were used as immunogen sequences to generate the antibody, GenBank no NP_775832.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 222643
Test Specificity This antibody is reactive to Chimpanzee, Human.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.