Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC5D/UNC5H4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981414
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1414 Supplier Bioss Supplier No. BS11494R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC5D, also named as KIAA1777 and UNC5H4, belongs to the unc-5 family. It is a receptor for netrin. It may be involved in axon guidance by mediating axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand.

Specifications

Antigen UNC5D/UNC5H4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UNC5D
Gene Accession No. Q6UXZ4
Gene Alias D930029E11Rik; FLJ16019; KIAA1777; KIAA1777Unc5h4; mKIAA1777; Netrin receptor UNC5D; netrin receptor Unc5h4; PRO34692; Protein unc-5 homolog 4; Protein unc-5 homolog D; unc5 homolog 4; unc-5 homolog 4; unc-5 homolog D; unc-5 homolog D (C. elegans); unc-5 netrin receptor D; UNC5D; Unc5h4; UNQ6012/PRO34692
Gene Symbols UNC5D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UNC5H4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137970
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.