Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC5H1/Unc5a Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300125PCP
Description
UNC5H1/Unc5a Polyclonal antibody specifically detects UNC5H1/Unc5a in Human samples. It is validated for ELISASpecifications
UNC5H1/Unc5a | |
Polyclonal | |
ELISA | |
KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5H1FLJ16449 | |
Produced in rabbits immunized with purified, recombinant Human UNC5H1/Unc5a (Accession#: NP_588610.2; Met1-Tyr306) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
90249 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction