Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC5H3/UNC5C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981413
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1413 Supplier Bioss Supplier No. BS11493R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin -like domains and 2 type I thrombospondin motifs in the extracellular region.

Specifications

Antigen UNC5H3/UNC5C
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Unc5c
Gene Accession No. O95185
Gene Alias B130051O18Rik; Netrin receptor UNC5C; Protein unc-5 homolog 3; Protein unc-5 homolog C; Rcm; Rostral cerebellar malformation protein; unc5 (C.elegans homolog) c; unc5 homolog 3; unc-5 homolog 3; unc-5 homolog C; unc-5 homolog C (C. elegans); unc-5 netrin receptor C; UNC5C; UNC5H3
Gene Symbols Unc5c
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UNC5H3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8633
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.