Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC79 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584881 Supplier Invitrogen™ Supplier No. PA584881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFALPEMSLD DHPDPGTEGE KPGELMPSSG AKTVLLKVPE DAENPTESEK PDTSAESDTE QNPERKVEED GAEESEFKIQ IVPRQRKQRK IAVSA Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 80%.

The NALCN channel is responsible for Na(+) leak currents. The protein encoded by this gene, along with UNC80, is an accessory subunit of the NALCN channel that contributes to the Ca(2+) sensitivity of the channel. [provided by RefSeq, Sep 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC79
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UNC79
Gene Accession No. Q9P2D8
Gene Alias KIAA1409; protein unc-79 homolog; UNC79; unc-79 homolog (C. elegans)
Gene Symbols UNC79
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNC79. Recombinant protein control fragment (Product #RP-108591).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.