Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UNC80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113286
Description
UNC80 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| UNC80 | |
| Polyclonal | |
| Unconjugated | |
| UNC80 | |
| C030018G13Rik; C230061B10Rik; C2orf21; Kiaa1843; LOC689991; mUNC-80; NALCN channel associated protein; protein unc-80 homolog; similar to CG18437-PA; UNC80; UNC-80; unc-80 homolog; unc-80 homolog (C. elegans); unc-80 homolog, NALCN activator; WUGSC:H_NH0436C12.1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 285175 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N2C7 | |
| UNC80 | |
| Recombinant Human Protein unc-80 homolog protein (124-388AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction