Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC84A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552564 Supplier Invitrogen™ Supplier No. PA552564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGNFFSFLPV LNWASMHRTQ RVDDPQDVFK PTTSRLKQPL QGDSEAFPWH WMSGVEQQVA SLSGQCHHHG ENLRELTTLL QKLQARVDQM EGGAAGPSAS VRDAVGQPPR ETDFMAFHQE HEVRMSHLED ILGKLR Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.

This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC84A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUN1
Gene Accession No. O94901
Gene Alias 4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a
Gene Symbols SUN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNC84A. Recombinant protein control fragment (Product #RP-89514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23353
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.