Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC84A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598345 Supplier Invitrogen™ Supplier No. PA598345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC84A Polyclonal Antibody for Western Blot, ELISA

This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC84A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias 4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana SUN domain-containing protein 1 protein (2-105AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.