Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC84B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA542981

Catalog No. PIPA542981


Only null left
Add to cart

Description

Description

Peptide sequence: SRRPDEGWEA RDSSPHFQAE QRVMSRVHSL ERRLEALAAE FSSNWQKEAM Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%.

UNC84B may form a physical interaction between the nuclear envelope and the centrosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UNC84B
Polyclonal
Unconjugated
SUN2
B230369L08Rik; C030011B15; FRIGG; KIAA0668; nuclear envelope protein; protein unc-84 homolog B; Rab5-interacting protein; rab5IP; RGD1563141; Sad1 and UNC84 domain containing 2; Sad1 unc-84 domain protein 2; sad1/unc-84 protein-like 2; SUN domain-containing protein 2; SUN2; unc-84 homolog B; UNC84B
Rabbit
Affinity chromatography
RUO
25777
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UH99
SUN2
Synthetic peptide directed towards the N-terminal of human UNC84B (aa 254-303).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.