Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC84B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551539 Supplier Invitrogen™ Supplier No. PA551539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKSEWQSMTQ ESFQESSVKE LRRLEDQLAG LQQELAALAL KQSSVAEEVG LLPQQIQAVR DDVESQFPAW ISQFLARGGG GRVGLLQREE MQAQLRELES KILTHVAEMQ GKSAREAAAS LSLTLQKEGV IGVTEEQVHH IVKQALQRYS E Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

UNC84B may form a physical interaction between the nuclear envelope and the centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC84B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUN2
Gene Accession No. Q9UH99
Gene Alias B230369L08Rik; C030011B15; FRIGG; KIAA0668; nuclear envelope protein; protein unc-84 homolog B; Rab5-interacting protein; rab5IP; RGD1563141; Sad1 and UNC84 domain containing 2; Sad1 unc-84 domain protein 2; sad1/unc-84 protein-like 2; SUN domain-containing protein 2; SUN2; unc-84 homolog B; UNC84B
Gene Symbols SUN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNC84B. Recombinant protein control fragment (Product #RP-91354).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.