Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC84B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595741 Supplier Invitrogen™ Supplier No. PA595741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, U-251MG, Mouse heart.

UNC84B may form a physical interaction between the nuclear envelope and the centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC84B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SUN2
Gene Accession No. Q8BJS4, Q9UH99
Gene Alias B230369L08Rik; C030011B15; FRIGG; KIAA0668; nuclear envelope protein; protein unc-84 homolog B; Rab5-interacting protein; rab5IP; RGD1563141; Sad1 and UNC84 domain containing 2; Sad1 unc-84 domain protein 2; sad1/unc-84 protein-like 2; SUN domain-containing protein 2; SUN2; unc-84 homolog B; UNC84B
Gene Symbols SUN2
Host Species Rabbit
Immunogen Recombinant protein of human SUN2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223697, 25777, 315135
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.