Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC93A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520509
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520509 Supplier Invitrogen™ Supplier No. PA520509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-20509 can be used with blocking peptide PEP-0629.

Unc93a, a human homolog of the C. elegans Unc93 gene, was initially identified by the sequencing of chromosome 6q27 at D6S193 as loss at this region correlated with sporadic ovarian cancer. Further analysis however, showed no tumor suppressor role for Unc93a. Unlike Unc93b, another homolog of the C. elegans Unc93 gene that is crucial for Toll-like receptor (TLR) signaling, the function of Unc93a is unknown. It is expressed in testis, small intestine, spleen, prostate and ovary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC93A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene UNC93A
Gene Accession No. Q86WB7
Gene Alias dJ366N23.1; dJ366N23.1 (putative C. elegans UNC-93 (protein 1, C46F11.1) LIKE); dJ366N23.2; dJ366N23.2 (putative C. elegans UNC-93 (protein 1, C46F11.1) C-terminal LIKE); HmUnc-93A; Protein unc-93 homolog A; unc93 homolog A; unc-93 homolog A (C. elegans); UNC93A; Unc-93A
Gene Symbols UNC93A
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the carboxy terminus of human Unc93a.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54346
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.