Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC93B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558510 Supplier Invitrogen™ Supplier No. PA558510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NHYLYDLNHT LYNVQSCGTN SHGILSGFNK TVLRTLPRSG N Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

The endoplasmic reticulum (ER) protein Unc93b, a human homolog of the C. elegans Unc93 gene, was initially identified by a forward genetic screen using N-ethyl-N-nitrosourea where a histidine-to-arginine substitution in Unc93b caused defects in Toll-like receptor (TLR) 3, 7 and 9 signaling. Unlike Unc93a, another homolog of the C. elegans Unc93 gene whose function is unknown, Unc93b specifically interacts with TLR3, 7 and 9; the histidine-to-arginine point mutation used to identify Unc93b abolishes this interaction. Mice carrying this point mutation are highly susceptible to infection with a number of viruses, indicating that Unc93b plays an important role in innate immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC93B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UNC93B1
Gene Accession No. Q9H1C4
Gene Alias hUNC93B1; IIAE1; Protein unc-93 homolog B1; UNC93; unc93 homolog B; unc-93 homolog B; unc93 homolog B1; unc-93 homolog B1 (C. elegans); unc-93 related protein; UNC93B; Unc93b1; Unc-93B1
Gene Symbols UNC93B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNC93B1. Recombinant protein control fragment (Product #RP-97283).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.