Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC93B1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520510

Catalog No. PIPA520510


Only null left
Add to Cart

Description

Description

A suggested positive control is human heart tissue lysate. PA5-20510 can be used with blocking peptide PEP-0630.

The endoplasmic reticulum (ER) protein Unc93b, a human homolog of the C. elegans Unc93 gene, was initially identified by a forward genetic screen using N-ethyl-N-nitrosourea where a histidine-to-arginine substitution in Unc93b caused defects in Toll-like receptor (TLR) 3, 7 and 9 signaling. Unlike Unc93a, another homolog of the C. elegans Unc93 gene whose function is unknown, Unc93b specifically interacts with TLR3, 7 and 9; the histidine-to-arginine point mutation used to identify Unc93b abolishes this interaction. Mice carrying this point mutation are highly susceptible to infection with a number of viruses, indicating that Unc93b plays an important role in innate immunity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UNC93B1
Polyclonal
Unconjugated
UNC93B1
hUNC93B1; IIAE1; Protein unc-93 homolog B1; UNC93; unc93 homolog B; unc-93 homolog B; unc93 homolog B1; unc-93 homolog B1 (C. elegans); unc-93 related protein; UNC93B; Unc93b1; Unc-93B1
Rabbit
Antigen Affinity Chromatography
RUO
361689, 54445, 81622
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q8VCW4, Q9H1C4
UNC93B1
A 19 amino acid peptide from near the amino terminus of human Unc93b.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.