Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNCX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569485 Supplier Invitrogen™ Supplier No. PA569485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%; Rat: 79%; Zebrafish: 77%.

Transcription factor involved in somitogenesis and neurogenesis. Required for the maintenance and differentiation of particular elements of the axial skeleton. May act upstream of PAX9. Plays a role in controlling the development of connections of hypothalamic neurons to pituitary elements, allowing central neurons to reach the peripheral blood circulation and to deliver hormones for control of peripheral functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNCX
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UNCX
Gene Accession No. A6NJT0
Gene Alias Chx4; Homeobox protein unc-4 homolog; homeobox protein unc-4 homolog; LOW QUALITY PROTEIN: homeobox protein unc-4 homolog; homeobox protein Uncx4.1; LOW QUALITY PROTEIN: homeobox protein unc-4 homolog; paired-type homeodomain transcription factor 1; Phd1; truncated UNCX homeobox protein; UNC homeobox; UNC homeobox protein; Unc4.1 homeobox; Uncx; Uncx4.1; UNCX41
Gene Symbols UNCX
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human UNCX.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340260
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.