Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecafluoropentyl Iodide (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. U008125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U008125G 25 g
U00815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U008125G Supplier TCI America Supplier No. U008125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 638-79-9
Molecular Formula C5F11I
Molecular Weight (g/mol) 395.942
MDL Number MFCD08461629
InChI Key KCEJJSGJNCSQFI-UHFFFAOYSA-N
Synonym Perfluoropentyl Iodide
PubChem CID 69494
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5-undecafluoro-5-iodopentane
SMILES C(C(C(F)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 94°C
Quantity 25 g
Formula Weight 395.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecafluoropentyl Iodide (stabilized with Copper chip)
EINECSNumber (2)-0090
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.