Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma-Undecalactone, 98%

Catalog No. p-3779866
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 104-67-6 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-67-6
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00005405
InChI Key PHXATPHONSXBIL-UHFFFAOYSA-N
Synonym gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone
PubChem CID 7714
IUPAC Name 5-heptyloxolan-2-one
SMILES CCCCCCCC1CCC(=O)O1

Specifications

Color Colorless
Density 0.9425g/mL
Boiling Point 286.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 25 mL
Specific Gravity 0.9425
Formula Weight 184.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material γ-Undecalactone
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-225-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.