Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanoic acid, 98%

Catalog No. p-7081323
Click to view available options
Quantity:
50 g
250 g

CAS: 112-37-8 | C11H22O2 | 186.295 g/mol

Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-37-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00002730
InChI Key ZDPHROOEEOARMN-UHFFFAOYSA-N
Synonym hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
PubChem CID 8180
ChEBI CHEBI:32368
IUPAC Name undecanoic acid
SMILES CCCCCCCCCCC(=O)O

Specifications

Melting Point 26°C to 29°C
Boiling Point 283°C to 285°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 1759287
Solubility Information Insoluble in water.
Formula Weight 186.3
Percent Purity 98%
Chemical Name or Material Undecanoic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c
EINECSNumber 203-964-2
RTECSNumber YQ2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.