Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanoic acid, 99%

Catalog No. AC173970250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 112-37-8 | C11H22O2 | 186.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-37-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.29
MDL Number MFCD00002730
InChI Key ZDPHROOEEOARMN-UHFFFAOYSA-N
Synonym hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
PubChem CID 8180
ChEBI CHEBI:32368
IUPAC Name undecanoic acid
SMILES CCCCCCCCCCC(=O)O

Specifications

Melting Point 28.5°C
Color White to Yellow
Boiling Point 284.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9CO2H
Quantity 25 g
Beilstein 02, 358
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,cresols and 1-butanol
Formula Weight 186.29
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Undecanoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-964-2
RTECSNumber YQ2275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.