Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecylbenzene 98.0+%, TCI America™
SDP

Catalog No. U001025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U001025ML 25 mL
U00105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U001025ML Supplier TCI America Supplier No. U001025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6742-54-7
Molecular Formula C17H28
Molecular Weight (g/mol) 232.411
MDL Number MFCD00026549
InChI Key XBEADGFTLHRJRB-UHFFFAOYSA-N
Synonym 1-phenylundecane, benzene, undecyl, n-undecylbenzene, undecanylbenzene, undecane, 1-phenyl, alkyl c10-c13 benzene, unii-8c9lpo3q9t, benzene, c10-13-alkyl derivs., benzene, mono-c10-13-alkyl derivs., 8c9lpo3q9t
PubChem CID 23194
IUPAC Name undecylbenzene
SMILES CCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 232.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecylbenzene
EINECSNumber (3)-0021
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.