Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNG Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149985AF647
Description
UNG Polyclonal antibody specifically detects UNG in Human samples. It is validated for Western BlotSpecifications
UNG | |
Polyclonal | |
Western Blot | |
DGUEC 3.2.2, HIGM4EC 3.2.2.27, UDGDKFZp781L1143, UNG15, UNG1uracil-DNA glycosylase 1, uracil-DNA glycosylase 2, UNG2, uracil-DNA glycosylase | |
The immunogen for this product maps to a region between residue 263 and 313 of human Uracil-DNA Glycosylase using the numbering given in entry NP_550433.1 (GeneID 7374). | |
0.1 mL | |
Base Excision Repair, Cancer, DNA Repair, Stem Cell Markers | |
7374 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction