Learn More
UNG Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302888AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The human UNG gene encodes both nuclear (UNG2) and mitochondrial (UNG1) forms of uracil-DNA glycosylase. These forms are generated by alternative splicing and the use of two differentially regulated promoters, PA and PB (Slupphaug et al., 1993; Nilsen et al., 1997) The cDNAs for UNG1 and UNG2 are of similar size (2061 and 2058 bp, respectively) and as a result the corresponding mRNAs are not resolved as two species in gel electrophoresis (Huag et al., 1998). Nuclear UNG2 differs from mitochondrial UNG1 in 44 amino acids of the N-terminal sequence that is not necessary for catalytic activity. A major role of the UNG gene products is to repair mutagenic U:G mispairs caused by cytosine deamination. For example, UNG2 removes misincorporated dUMP residues. The level and expression pattern of UNG1 and UNG2 differs between cell and tissue type (Huag et al. 1998). Additionally, the expression of UNG is cell cycle regulated (Nagelhus et al. 1995).The expression and activity of UNG has been found in general to be higher in proliferating as compared to nonproliferating tissues and cells (reviewed in Kruman et al. 2004). UNG1 is a 304 amino acid protein. UNG2 is a 313 amino acid protein.Specifications
UNG | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P13051 | |
UNG | |
Between 263 and 313 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UNG | |
DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7374 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.