Learn More
Owen Mumford™ Unifine™ SafeControl™ Safety Pen Needle
SDP
Catalog No. 22024109 Shop All Owen Mumford, Inc. Products
Click to view available options
Length (Metric):
5 mm
8 mm
This item is not returnable. View return policy
Catalog No. 22-024-109
Supplier: Owen Mumford™ AN 7935
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The next generation safety pen needle providing both safety and control featuring dual-protection on both ends, protecting both the patient and healthcare professional, and a visible needle to help prevent premature activation and provide confidence in dose-delivery.
  • Intuitive design makes operation easy
  • Visible needle utilizes the same technique as conventional pen needles
  • Innovative push tab for one-handed activation of the safety mechanism
  • Green cap for sterility and protection from the needle prior to injection
  • Safety guard that covers the needle after injection to help prevent accidental needle sticks
  • Dual safety indicator with visual and audible confirmation
  • Both ends of the needle covered to protect both patient and healthcare professional
  • Universal fit-compatible with most injection pens
Specifications
Autoclavable Not Autoclavable
Needle Gauge 30 G
For Use With (Application) Drug Delivery Using Injection Pens
Length (Metric) 8 mm
Disposable Single-use
Product Line Unifine SafeControl
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.