Learn More
Protective Industrial Products Uniform Technology™ Altessa Grid Navy Frock
Catalog No. 17010672
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Reusable cleanroom frock features raglan sleeves and ESD knit cuffs for use in ISO 5 or higher compatible environments.
  • Navy
  • Fabric 99% Polyester 1% Carbon
  • ISO 5 (Class 100)
  • Features raglan sleeve, military collar, coil zipper at front opening, snaps at collar and snap 2 in. below bottom of zipper, sleeves with ESD knit cuff
Specifications
Product Type Controlled Environments Frock
Cuff Style ESD Knit
For Use With (Application) Electronics and Aerospace, Food Processing, Controlled Environments, Film Coating and Imaging Manufacturing
Collar Type Military
Autoclavable Autoclavable
Cleanroom Class Class 100 (ISO 5)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.