Learn More
Thermo Scientific™ Uniguard™ Direct-Connection Guard Cartridge Holders
Catalog No. 03052393
Change view
Click to view available options
For Use With (Equipment):
1 mm I.D. Guard Columns
2.1 to 3 mm I.D. Guard Columns
4.6 mm I.D. Guard Columns
Waters Columns Replacement Tips
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-052-393 Waters Columns Replacement Tips
85000 4.6 mm I.D. Guard Columns
85100 1 mm I.D. Guard Columns
85200 2.1 to 3 mm I.D. Guard Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-052-393 Supplier Thermo Scientific™ Supplier No. 850WT
Add to Cart
Add to Cart
Eliminate the requirement for extra fittings with Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders.
Eliminate the requirement for extra fittings using Thermo Scientific™ Uniguard Direct-Connection Guard Cartridge Holders. They are reusable, stainless-steel guard cartridge holders that attach directly to the analytical column inlet.
  • With PEEK 0.0625in. male outlet that fits many columns
  • 0.0625in. female inlet tip can be used with various standard fittings
Ordering Information:
Other column dimensions are available. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Description Guard Cartridge Holder for Waters Columns Replacement Tips
For Use With (Equipment) Waters Columns Replacement Tips
Type Cartridge Holder
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.