Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ Union Titanium 1/16 in. Female to 1/16 in. Female Chromatography Fitting

Catalog No. 01257016 Shop All Cytiva Products
Change view
Click to view available options
Thread Size:
1/16 in. (f)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
01-257-016 1/16 in. (f)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-016 Supplier Cytiva™ Supplier No. 18385501
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with ÄKTA™ systems and tubing connectors with 1/16 in. outer diameter.

  • Use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant
  • Use together with Fingertight connector 1/16 in. (m) 18111255 or Fingertight connector 1/16 in. (m) narrow 18117263
TRUSTED_SUSTAINABILITY

Specifications

Type Fitting
Color Silver
Thread Size 1/16 in. (f)
Material Titanium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.