Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Computype Unique Device Identification (UDI) Medical Device Labeling Assessment
SDP

Catalog No. CMPUDIASSMT
Change view
Click to view available options
Product Type:
Workplace Safety Compliance Assessments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
CMPUDIASSMT Workplace Safety Compliance Assessments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
FDA 21CFR UDI
Catalog No. CMPUDIASSMT Supplier Computype Supplier No. CMPUDIASSMT

To request a quote for this item, please select Request a Quote to reach our dedicated support team.

FDA 21CFR UDI

Our comprehensive assessment addresses FDA 21CFR UDI requirments by reviewing best practices to support accurate and efficient medical device identification.

The FDA established the Unique Device Identification system to identify medical devices sold in the United States from manufacturing through distribution to patient use. Computype can assess your new product requirements or existing way of work for workflow enhancements. There are several ways to uniquely identify a device, and Computype has experts that can advise on which options will work for your specific application so you are not only in compliance but following best practices.

  • Bar code printing, grading and scanning expertise that ensures performance and compliance.
  • On-demand labeling options
  • Preprinted label options
  • Sequence management
  • Durable materials that withstand freezing, thawing, and chemical exposure to ensure your device identification.
  • Cryogenic environments
  • Nase free environments
  • Extreme heat exposure
  • Sterilization ready materials
  • Environmental friendly sustainable label materials
  • Label printer support and recommendations and optimization with your workflow in mind.
  • Bar code label printing software and associated design programming
  • Color coding for clarification and workflow efficiency
  • Automatic labeling automationLabware Prep - outsource the labeling of components and tubes for ultimate convenience and accuracy
  • Direct mark ink-jet
  • Laser marking

Specifications

Product Type Workplace Safety Compliance Assessments
Federal Regulation FDA 21 CFR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.