Learn More
Simport™ Scientific UNISETTE™ Tissue Processing/Embedding Cassette
Shop All Simport Scientific ProductsDescription
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
| Writing Face Angle | 35° |
| Color | Orange |
| Material | Acetal |
| Product Type | Tissue Processing/Embedding Cassette |
For in vitro diagnostic (IVD) use only.