Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks for Primera Printers
Catalog No. 22038002 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-002 Pink
22-038-001 White
22-038-003 Green
22-038-004 Yellow
22-038-006 Peach
22-038-007 Tan
22-038-008 Gray
22-038-009 Lilac
22-038-010 Orange
22-038-011 Aqua
22-038-005 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-038-002 Supplier Simport™ Scientific Supplier No. M4053T
Add to Cart
Add to Cart
Secure specimens during processing with Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks for Primera Printers. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
  • In Quickload™ stacks, for Primera printers
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 1000/Pk.
Color Pink
Material Acetal