Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Owen Mumford™ Unistik™ Pro Single Use Safety Lancet
SDP

Catalog No. 22024104
Click to view available options
Flow Type:
High
Low
Medium
Needle Gauge:
21 G
25 G
28 G
This item is not returnable. View return policy
This item is not returnable. View return policy

Featuring a syringe-like grip and a push-button design, Unistik™ Pro offers a reliable blood sampling solution that’s ergonomic, intuitive and affordable.

  • Comfort Zone™ Technology for enhanced patient comfort
  • Intuitive design makes blood sampling easy for healthcare professionals
  • Needle is hidden before use and permanently retracts after use to help protect against accidental needle sticks and cross contamination

Specifications

Disposable Single-use
Style Top Activated
For Use With (Application) Capillary Blood Sampling
Packaging Quantity 200/Pk.
Sterility Sterile
Color Blue
Product Type Unistik™ Pro Safety Lancet
Needle Gauge 28 G
Flow Type Low
Depth (Metric) 1.2 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.