Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Sterile Media Bottles, PETG
SDPSCI_ED

Catalog No. S80028 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S80028 250 mL
S80022 30 mL
S80027 60 mL
S80025 125 mL
S80021 500 mL
S80026 1000 mL
S80029 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S80028 Supplier United Scientific™ Supplier No. BMPETG250
Add to Cart
Add to Cart

Store and transport your liquids with leakproof, freezer safe UniStore PETG Media Bottles.

  • UniStore™ polyethylene terephthalate glycol (PETG) bottles are designed for long-term storage of media, biological buffers, and various reagents.
  • Bottles are leakproof (tested to 95 kPa), durable, as well as freezer-safe, making them ideal for storage and transport.
  • Includes a high-density polyethylene (HDPE) closure with heat-shrink band for a tamper evident seal.
  • Bottles feature raised graduations for easy readability.
  • Bottles are suitable for frozen storage to -40°C
  • Bottles are Gamma sterilized and packaged in shrink-wrapped trays within an outer bag.
  • PETG bottle with HDPE closure.
  • Manufactured in ISO-8 Cleanroom (100K).
  • Bottle materials meet USP Class VI requirements and are noncytotoxic, nonhemolytic, nonpyrogenic.
  • Gamma sterilized and have Sterility Assurance Level (SAL) of 10-6.
  • Temperature range: -40° to 70°C
  • Bottles are leakproof (tested to 95 kPa) and have a heat-shrink band around closure for tamper-evident seal.
  • Size range from 30 mL to 2000 mL

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.5 oz.
Color Clear
Material PETG
Quantity 48
Sterility Sterile
Closure Included Yes
Disposable Single-use
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 30 cm
Product Type Media Bottle
For Use With (Equipment) Mixing, sampling, and storage applications
Width (English) 10.25 in.
Length (English) 15 in.
Height (English) 11.8 in.
Closure Material HDPE
Closure Size 28.5 mm
Closure Type Tamper-evident Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.