Learn More
United Scientific Supplies Clear Base Vacuum Desiccators
SDPSCI_ED
Made of polypropylene (PP) and polycarbonate (PC).
Supplier:  United Scientific Supplies 55216
 View more versions of this product
Catalog No. S13077
Only null left
Add to Cart
Description
Description
  • Can hold vacuum up to 740mmHg for 24 hours without any greasing
  • Autoclavable at 120°C for 15 minutes
  • Top dome, molded in rigid and transparent PC, gives crystal-clear view of sample
  • Knurled knob on top provides easy handling of dome
  • Transparent PC bottom provides increased viewing clarity
  • Internal groove on flange holds silicon rubber O-ring
  • PP stopcock has self-lubricating PTFE plug
  • Three-way plug opens to create vacuum, closes to hold vacuum, and opens again to release vacuum
Specifications
Specifications
Desiccator
254 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.