Learn More
United Scientific Supplies United Scientific™ Stainless Steel Forceps, 5 in.
SDP
Catalog No. 01672032 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Tip Style:
Blunt
Sharp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
01-672-032 Sharp
01-672-029 Blunt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-672-032 Supplier United Scientific Supplies Supplier No. FOMF05
Add to Cart
Add to Cart
Forceps support general laboratory and dissection handling.
Good quality stainless steel forceps for general use in the laboratory. All feature serrated tips. Non-sterile, autoclavable.
  • Stainless steel forceps for general lab use
  • Serrated tips for secure handling
  • Non-sterile and autoclavable
Specifications
Tip Style Sharp
Length (English) 5 in.
Material Stainless Steel
Product Type Forceps
Type Stainless Steel Sharp Forceps
Sterility Non-sterile
Style Standard