Learn More
United Scientific Supplies Color Mixing Apparatus
Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Light projector; With power supply and manual
Description
- Compact and economical
- The projector mounting allows the beam angles to be adjusted to vary the overlap of the three colored images
- The intensity of each LED can be adjusted separately by a potentiometer to give a convincing white light or various shades of mixed colors
- Colored shadows can also be explored in a darkened room
Specifications
Specifications
| Product Type | Color Mixing Apparatus |
| Includes | A wall mount power supply and manual, red, green and blue color light projector |