Learn More
Unity Lab Services™ Pipette Support Plans
Fast, quality-minded service that meets your diverse needs
$55.00 - $124.00
Specifications
| Service Offering | Pipette Validation |
|---|---|
| Includes | Thorough external pipette inspection; Gravimetric validation to determine if desired accuracy and precision is achieved |
| Catalog Number | Mfr. No. | Calibration Points | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Calibration Points | For Use With (Equipment) | Price | Quantity & Availability | ||||
12-102-105
![]() |
Unity Lab Services™
VFC2X10DSC |
2 x 10 Measurements | Single-channel Pipette |
Each for $85.00
|
|
||||
12-102-103
![]() |
Unity Lab Services™
VFC2X5DSC |
2 x 5 Measurements | Single-channel Pipette |
Each for $61.25
|
|
||||
12-102-104
![]() |
Unity Lab Services™
VFC2X5DMC |
2 x 5 Measurements | Multi-channel Pipette |
Each for $121.00
|
|
||||
12-102-106
![]() |
Unity Lab Services™
VFC2X10DMC |
2 x 10 Measurements | Multi-channel Pipette |
Each for $124.00
|
|
||||
12-102-111
![]() |
Unity Lab Services™
ATV1 |
One Additional Test Volume | Single or Multi-channel Pipette |
Each for $55.00
|
|
||||
Description
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
- Service for any pipette brand and model
- Reliable service options by our highly trained technicians
- Fast and convenient prepaid pipette shipping packages
Basic – Validation-Fast Check
The Validation-Fast Check Plan combines a thorough external pipette inspection with gravimetric validation to determine if desired accuracy and precision is achieved. This plan includes a validation pass/fail report.
Ideal for customers who require:
- An interim validation check that occurs in-between routine scheduled preventive maintenance and calibration
- Repairs for dropped/disassembled pipettes or want to make sure pipettes are performing as desired
- A cost-friendly service option
† If a pipette fails Validation-Fast Check, the customer can request to be contacted with a recommended service course of action (i.e. GxP Preventive Maintenance and Calibration Plan.)
Specifications
| Pipette Validation | |
| Thorough external pipette inspection; Gravimetric validation to determine if desired accuracy and precision is achieved |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.