Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Universal 7 Pipette Work Station, Model 337
SDP

Catalog No. p-7111682
Click to view available options
Color:
Ice Blue
Mint Green
Pastel Rose
Vanilla Yellow

Keep your workstation organized with this universal pipette stand for up to 7 pipettes. Available in a variety of colors.

  • Space-saving pipette stand holds up to seven single-channel pipettes of most brands
  • Made of sturdy polyamid material, easy to clean
  • Heavy bottom plate and anti-slip pads guarantee bench stability
  • The work station fits new and old generations of Socorex and other brand pipettes
TRUSTED_SUSTAINABILITY

Specifications

Includes Anti-slip pads
Product Type Pipette Work Station
Description Model 337
Color Ice Blue
Holds Pipettes (7 Single Channel)
Material Polyamide
Positions 7
Shape Round
For Use With (Equipment) Univeral
No. of Racks 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.