Learn More
Eppendorf™ Universal Buckets for S-4xUniversal Rotor
Click to view available options
Quantity:
2 Pk.
4 Pk.
Description
- Suitable for S-4xUniversal Rotor
- Accepts adapters for tubes, flasks, and plates
- Variable places depending on adapter used
Specifications
Specifications
For Use With (Equipment) | S-4xUniversal Rotor |
Quantity | 2 Pk. |