Learn More
R&D Systems™ Universal Colorimetric PARP Assay Kit - Histone-Coated Wells
GSA_VA
Catalog No. 4677096K
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
4677096K 96 Tests
1 options
This item is not returnable. View return policy
Catalog No. 4677096K Supplier R&D Systems™ Supplier No. 4677096K
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
PARP-HSA, 10 units/uL, 50uL; Histone-Coated Clear Strip wells, 96 wells; 200mM 3-Aminobenzamide, 60uL; Strep-HRP, 30uL; TACS-Sapphire™, 10mL; 10X Activated DNA, 300uL; Buffer, Diluent and PARP Cocktail
Measures the activity of Poly (ADP-ribose) Polymerase (PARP) in cells and tissues by detecting the incorporation of biotinylated Poly (ADP-ribose) (PAR) onto histone proteins coating a 96-stripwell microplate
This assay is useful for testing whether DNA is damaged, if damaged DNA is from non-apoptotic cells, or the effectiveness of PARP inhibitors
Non-radioactive
Amenable to high-throughput
Highly sensitive: capable of measuring 0.01 Units of PARP/well
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 96 Tests
Content And Storage The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date.