Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics™ Universal Fit Pipette Tip

Catalog No. 23444003
Click to view available options
Volume (Metric):
10 μL
100 μL
1250 μL
200 μL

Great Lakes Diagnostics™ universal fit pipette tips are available in different volumes and lengths. Compatible with a broad range of pipettes.

  • Sterile
  • Filtered
  • Racked
  • DNA/RNA free
  • Max Recovery
  • Compatible with:
    • Eppendorf Reference™ SC, vol .5-10 μL, 2-20 μL, 10-100 μL, 50-200 μL, 100-1000 μL
    • Thermo Finnpipette™ F1/F2 SC, 1-10 μL, 2-20 μL, 10-100 μL, 20-200 μL, 100-1000 μL
    • Finnpipette F3 SC 1-10 μL, 10-100 μL
    • Biohit Proline™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
    • Gilson Pipetman™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
    • Axygen™ SC and MC pipettes.
    • Corning™ pipettes.

Specifications

Volume (Metric) 100 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Disposable Single-use
For Use With (Equipment) Axygen™ SC and MC, Biohit™ Proline™ SC, Corning™, Eppendorf Reference, Gilson™ Pipetman™ SC, Axygen™ SC and MC, Corning™
Graduated No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.