Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Universal Indicator, pH 2 - 10, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Specifications
Specifications
Chemical Name or Material | Universal Indicator |
Name Note | pH 2 - 10 |
CAS | 64-17-5, 67-56-1, 62625-21-2, 34722-90-2, 60-11-7, 493-52-7 |
CAS Min % | 98.0, 0.12, 0.1, 0.07, 0.05, 0.02 |
CAS Max % | 100.0, 0.13, 0.1, 0.08, 0.05, 0.03 |
Grade | Indicator |
Packaging | Amber Glass Bottle |
Color | Dark Brown |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction