Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safetec™ Universal Precaution Kit
SDP

Catalog No. p-37104
Encompass
Click to view available options
Type:
Universal Precaution Compliance Kit (hard case)
Universal Precaution Compliance Kit (poly bag)

Catalog No. 19-314-633

Supplier: Safetec™ 17102

Only null left
Add to Cart
Add to Cart

Includes

  • One pair of vinyl gloves
  • Protective apron
  • Combo mask/safety shield
  • Red Z™ solidifier (10g. pouch)
  • Scoop/scraper
  • SaniZide Plus™ germicidal wipe (8 x 8 in.)
  • Red bio-hazard waste Bbg (24 x 24 in.)
  • Twist tie
  • p.a.w.s.™ antimicrobial hand wipe (5 x 8 in.)
  • Identification tag
  • Instructions

A safe and cost-effective system for the clean-up of potentially infectious and harmful blood and body fluid spills

Single-use kit combines personal protection and cleanup items mandated by OSHA, CDC, and State Health Departments to aid in the cleanup, transportation and disposal of infectious blood or body fluid spills.

For cleanup of potentially infectious and harmful blood and body fluid spills.

Order Info

Available in a durable hard case or a polybag container

Compliance

Contains items mandated by OSHA, CDC and state health departments.

Specifications

Certifications/Compliance OSHA, CDC
Type Universal Precaution Compliance Kit (hard case)
For Use With (Application) Clean-up, transportation and disposal of potentially infectious blood or body fluid spills
Includes One pair of vinyl gloves, protective apron, combo mask or safety shield, Red Z™ solidifier (10 g pouch), scoop or scraper, SaniZide Plus™ germicidal wipe (8 x 8 in.), red biohazard waste bag (24 x 24 in.), twist tie
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.