Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Y, Angled
SDP
Catalog No. 06717753 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Includes:
Deactivation
Siltek Deactivated
Undeactivated
Quantity:
1 Ea.
3 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
06-717-753 Undeactivated 3 Pk.
06-611-158 Undeactivated 1 Ea.
06-611-159 Deactivation 1 Ea.
06-611-162 Siltek Deactivated 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-717-753 Supplier Restek™ Supplier No. 20404
Connect analytical columns, guard columns, transfer lines or detectors
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Type Universal Angled Y Press-Tight Connector
Includes Undeactivated
Quantity 3 Pk.
Material Glass