Description
- Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
- Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
- Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Specifications
| Type | Universal Y Press-Tight Connector |
| Includes | Undeactivated |
| Quantity | 3 Pk. |