Learn More
Novus Biologicals™ Universal Reverse Triiodothyronine ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 93.75-6000 pg/mL |
| Assay Sensitivity | 56.25 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA, Competitive ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | 3,3',5'-T3, 3,3',5'-Triiodo-L-thyronine, 5817-39-0, Isoliothyronine, Reverse L-triiodothyronine, Reverse T3, rT(3) |
| Target | Universal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.