Learn More
Thermo Scientific™ Universal Sample Holders for Thermo Scientific Spectronic™ IR100 FT-IR Spectrometer
Description
Universal Sample Holders. Available in two sizes.
- Mounts for salt plates, pellets, or films
- Fit in standard slide-card mounts
Specifications
Specifications
| For Use With (Equipment) | Spectronic IR100 FT-IR spectrometer |
| Type | Universal Sample Holder |
| Diameter (Metric) | 13 mm |
| For Use With (Application) | For positioning of KBr pellets, thin films, or IR windows for transmission analyses |