Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hardy Diagnostics™ Universal Transport Medium

Click to view available options
Includes:
No swabs
Description
- Has been used successfully for Rapid Antigen Testing, DFA, Viral Culture and for Molecular-based Assays.
- Can be conveniently stored at room temperature prior to use.
- Selection of tube sizes and collection applicators available. Also available in bulk or as patient collection packs, which incorporate different combinations of Polyester swabs, as well as mini tip or regular flocked swabs (FLOQSwabs™) and media tubes to suit different work patterns and preferences.
- Safe and reliable molded breakpoint swab - All swabs provided with UTM™ feature either a pre-scored breakpoint (polyester swabs) or a unique molded breakpoint (flocked swabs). Pre-scored and molded breakpoints feature allows for safe, reliable breaking of the swab into transport tube.
- Shatterproof skirted tubes with distinctive internal conical shape enable centrifugation of samples and allows the tubes to stand upright on the laboratory bench.
- Three glass beads in each tube facilitate the release and dispersion of patient sample material and virus particles from the swab during vortexing.
Specifications
Specifications
Media Type | UTM-RT |
Sterility | Sterile |
Closure Color | Red |
Closure Type | Screw Cap |
Color | Light Orange-Red |
For Use With (Application) | Collection, transport, maintenance and long term freeze storage; Rapid Antigen Testing, DFA, Viral Culture and Molecular-based Assays. |
Includes | No swabs |
Quantity | 300 Pk. |
Transport Temperature | 2°C to 25° |
Tube Size | 16 x 100 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction