Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555737 Supplier Invitrogen™ Supplier No. PA555737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYKSTKCNDM QQSGSCPRGP FCAFAHVEQP PLSDDLQPSS AVSSPTQPGP VLYMPSAAGD SVPVSPSSPH APDLSALLCR NSSLGSPSNL CGSPPGSIRK PPNLEGIVFP GES Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.

Sequence-specific RNA-binding protein which plays an important role in the establishment and maintenance of the early morphology of cortical neurons during embryonic development. Acts as a translation repressor and controls a translationally regulated cell morphology program to ensure proper structuring of the nervous system. Translational control depends on recognition of its binding element within target mRNAs which consists of a mandatory UAG trimer upstream of a U/A-rich motif. Associated with polysomes (PubMed:25737280). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UNK
Gene Accession No. Q9C0B0
Gene Alias B230379M23Rik; KIAA1753; mKIAA1753; RING finger protein unkempt homolog; UNK; UNKEMPT; unkempt family zinc finger; unkempt homolog; unkempt homolog (Drosophila); Zc3h5; ZC3HDC5; Zinc finger CCCH domain-containing protein 5; zinc finger CCCH type containing 5; zinc finger CCCH type domain containing 5; zinc finger CCCH-type containing 5; zinc finger CCCH-type domain containing 5
Gene Symbols UNK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNK. Recombinant protein control fragment (Product #RP-93268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.