Learn More
UNK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304713AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.Specifications
UNK | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8BL48, Q9C0B0 | |
Unk | |
Between 760 and 810 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UNK | |
ZC3H5, ZC3HDC5, RING finger protein unkempt homolog, zinc finger CCCH domain-containing protein 5, zinc finger CCCH type domain containing 5, zinc finger CCCH-type containing 5, zinc finger CCCH-type domain containing 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
217331, 85451 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.